SY3H3D6818DB Synthonix Corporation

2-(Piperazin-2-yl)ethanol dihydrochloride

  • MDL number No MDL found
  • Cas Number : 5169-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43565.html

Safety Information

MSDS https://synthonix.com/43565.html
Availability details for
Quantity
  • Availability
  • Add to Cart