SY3H3D6818DE Synthonix Corporation

Pentafluoroanilinium Trifluoromethanesulfonate

  • Cas Number : 912823-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43584.html

Safety Information

MSDS https://synthonix.com/43584.html
Availability details for
Quantity
  • Availability
  • Add to Cart