SY3H3D6818E3 Synthonix Corporation

Potassium 4-nitro-2-sulfonatobenzoate

  • Cas Number : 5344-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43592.html

Safety Information

MSDS https://synthonix.com/43592.html
Availability details for
Quantity
  • Availability
  • Add to Cart