SY3H3D6818E9 Synthonix Corporation

(S)-1-(4-Pyridyl)ethylamine

  • Cas Number : 27854-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43607.html

Safety Information

MSDS https://synthonix.com/43607.html
Availability details for
Quantity
  • Availability
  • Add to Cart