SY3H3D6818F0 Synthonix Corporation

5-Isopropyl-1H-pyrazole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 92933-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43660.html

Safety Information

MSDS https://synthonix.com/43660.html
Availability details for
Quantity
  • Availability
  • Add to Cart