SY3H3D6818F1 Synthonix Corporation

(1-Isopropyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 2170881-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43668.html

Safety Information

MSDS https://synthonix.com/43668.html
Availability details for
Quantity
  • Availability
  • Add to Cart