SY3H3D6818FE Synthonix Corporation

3-(1-Pyrrolidyl)-1-propanol

  • MDL number No MDL found
  • Cas Number : 19748-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43909.html

Safety Information

MSDS https://synthonix.com/43909.html
Availability details for
Quantity
  • Availability
  • Add to Cart