SY3H3D681900 Synthonix Corporation

2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane

  • MDL number No MDL found
  • Cas Number : 94242-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43937.html

Safety Information

MSDS https://synthonix.com/43937.html
Availability details for
Quantity
  • Availability
  • Add to Cart