SY3H3D681905 Synthonix Corporation

Pyrido[2,3-b]pyrazin-3(4H)-one

  • MDL number No MDL found
  • Cas Number : 35252-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44866.html

Safety Information

MSDS https://synthonix.com/44866.html
Availability details for
Quantity
  • Availability
  • Add to Cart