SY3H3D681909 Synthonix Corporation

5H-Pyrrolo[2,3-b]pyrazine-6-carboxylic acid

  • MDL number MFCD22068895
  • Cas Number : 1016241-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44883.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44883.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart