SY3H3D68195E Synthonix Corporation

1H-Pyrrolo[3,2-c]pyridin-6-amine

  • MDL number No MDL found
  • Cas Number : 1000342-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/45727.html

Safety Information

MSDS https://synthonix.com/45727.html
Availability details for
Quantity
  • Availability
  • Add to Cart