SY3H3D68195F Synthonix Corporation

(5H-Pyrrolo[2,3-b]pyrazin-7-yl)methanol

  • MDL number No MDL found
  • Cas Number : 20322-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45739.html

Safety Information

MSDS https://synthonix.com/45739.html
Availability details for
Quantity
  • Availability
  • Add to Cart