SY3H3D681962 Synthonix Corporation

3-Pyridyloxyacetic acid

  • MDL number No MDL found
  • Cas Number : 86649-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45834.html

Safety Information

MSDS https://synthonix.com/45834.html
Availability details for
Quantity
  • Availability
  • Add to Cart