SY3H3D681963 Synthonix Corporation

2-(Pyridin-4-yloxy)acetic acid

  • MDL number No MDL found
  • Cas Number : 58530-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45840.html

Safety Information

MSDS https://synthonix.com/45840.html
Availability details for
Quantity
  • Availability
  • Add to Cart