SY3H3D681965 Synthonix Corporation

2-(Pyridin-3-yl)acetamide

  • MDL number No MDL found
  • Cas Number : 3724-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/45977.html

Safety Information

MSDS https://synthonix.com/45977.html
Availability details for
Quantity
  • Availability
  • Add to Cart