SY3H3D68196A Synthonix Corporation

(R)-1-(Pyridin-2-yl)ethanol

  • Cas Number : 27911-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46382.html

Safety Information

MSDS https://synthonix.com/46382.html
Availability details for
Quantity
  • Availability
  • Add to Cart