SY3H3D68196C Synthonix Corporation

3-(1H-Pyrrol-2-yl)propanoic acid

  • MDL number No MDL found
  • Cas Number : 408309-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/46404.html

Safety Information

MSDS https://synthonix.com/46404.html
Availability details for
Quantity
  • Availability
  • Add to Cart