SY3H3D68196E Synthonix Corporation

N-Phenylmethanesulfonamide

  • MDL number No MDL found
  • Cas Number : 1197-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46415.html

Safety Information

MSDS https://synthonix.com/46415.html
Availability details for
Quantity
  • Availability
  • Add to Cart