SY3H3D681970 Synthonix Corporation

2-(Pyridin-2-ylthio)ethanol

  • Cas Number : 77555-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46439.html

Safety Information

MSDS https://synthonix.com/46439.html
Availability details for
Quantity
  • Availability
  • Add to Cart