SY3H3D681973 Synthonix Corporation

2-(Piperidin-3-yl)acetic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 71985-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46485.html

Safety Information

MSDS https://synthonix.com/46485.html
Availability details for
Quantity
  • Availability
  • Add to Cart