SY3H3D68197B Synthonix Corporation

(R)-1-(Pyridin-2-yl)ethanamine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1352640-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46583.html

Safety Information

MSDS https://synthonix.com/46583.html
Availability details for
Quantity
  • Availability
  • Add to Cart