SY3H3D68197E Synthonix Corporation

2-(Pyridin-4-yloxy)ethanamine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 442126-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46588.html

Safety Information

MSDS https://synthonix.com/46588.html
Availability details for
Quantity
  • Availability
  • Add to Cart