SY3H3D681982 Synthonix Corporation

Potassium 2-ethylhexanoate

  • MDL number MFCD00045896
  • Cas Number : 3164-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Lump / Colour: White
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46623.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46623.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart