SY3H3D681990 Synthonix Corporation

3-Isopropoxypyridine

  • Cas Number : 88111-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46891.html

Safety Information

MSDS https://synthonix.com/46891.html
Availability details for
Quantity
  • Availability
  • Add to Cart