SY3H3D681991 Synthonix Corporation

2-(Pyridin-3-yl)propan-2-ol

  • MDL number No MDL found
  • Cas Number : 15031-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46904.html

Safety Information

MSDS https://synthonix.com/46904.html
Availability details for
Quantity
  • Availability
  • Add to Cart