SY3H3D681995 Synthonix Corporation

2-Isopropylpyridin-3-amine

  • MDL number No MDL found
  • Cas Number : 1369105-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46959.html

Safety Information

MSDS https://synthonix.com/46959.html
Availability details for
Quantity
  • Availability
  • Add to Cart