SY3H3D681998 Synthonix Corporation

3-(Pyridin-4-yl)propan-1-amine

  • MDL number No MDL found
  • Cas Number : 30532-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/46966.html

Safety Information

MSDS https://synthonix.com/46966.html
Availability details for
Quantity
  • Availability
  • Add to Cart