SY3H3D68199B Synthonix Corporation

2-(Piperazin-1-yl)pyrimidin-5-ol

  • MDL number No MDL found
  • Cas Number : 55745-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47006.html

Safety Information

MSDS https://synthonix.com/47006.html
Availability details for
Quantity
  • Availability
  • Add to Cart