SY3H3D6819A1 Synthonix Corporation

N2-Isopropylpyridine-2,3-diamine

  • MDL number No MDL found
  • Cas Number : 24188-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47054.html

Safety Information

MSDS https://synthonix.com/47054.html
Availability details for
Quantity
  • Availability
  • Add to Cart