SY3H3D6819A2 Synthonix Corporation

1-Isopropyl-4-piperidone

  • MDL number No MDL found
  • Cas Number : 5355-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47115.html

Safety Information

MSDS https://synthonix.com/47115.html
Availability details for
Quantity
  • Availability
  • Add to Cart