SY3H3D6819A3 Synthonix Corporation

1-Isopropylpiperidin-3-ol

  • Cas Number : 3554-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47169.html

Safety Information

MSDS https://synthonix.com/47169.html
Availability details for
Quantity
  • Availability
  • Add to Cart