SY3H3D6819A8 Synthonix Corporation

1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 25957-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47872.html

Safety Information

MSDS https://synthonix.com/47872.html
Availability details for
Quantity
  • Availability
  • Add to Cart