SY3H3D6819F6 Synthonix Corporation

3-(Pyridin-4-yl)isoxazol-5(4H)-one

  • Cas Number : 101084-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47886.html

Safety Information

MSDS https://synthonix.com/47886.html
Availability details for
Quantity
  • Availability
  • Add to Cart