SY3H3D6819F8 Synthonix Corporation

Pyrrolo[1,2-b]pyridazine-6-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 118768-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47888.html

Safety Information

MSDS https://synthonix.com/47888.html
Availability details for
Quantity
  • Availability
  • Add to Cart