SY3H3D6819F9 Synthonix Corporation

4-(Pyridin-4-yl)thiazole-2-thiol

  • MDL number No MDL found
  • Cas Number : 77168-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47910.html

Safety Information

MSDS https://synthonix.com/47910.html
Availability details for
Quantity
  • Availability
  • Add to Cart