SY3H3D6819FD Synthonix Corporation

4-Phenyl-4H-1,2,4-triazole

  • MDL number No MDL found
  • Cas Number : 16227-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48388.html

Safety Information

MSDS https://synthonix.com/48388.html
Availability details for
Quantity
  • Availability
  • Add to Cart