SY3H3D6819FF Synthonix Corporation

1-(1H-Pyrazolo[3,4-b]pyridin-3-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 889451-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48400.html

Safety Information

MSDS https://synthonix.com/48400.html
Availability details for
Quantity
  • Availability
  • Add to Cart