SY3H3D681A03 Synthonix Corporation

5-(Pyridin-4-yl)thiazol-2-amine

  • MDL number No MDL found
  • Cas Number : 40353-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48422.html

Safety Information

MSDS https://synthonix.com/48422.html
Availability details for
Quantity
  • Availability
  • Add to Cart