SY3H3D681A05 Synthonix Corporation

3-(2-pyridyl)acrylicacid

  • MDL number No MDL found
  • Cas Number : 7340-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48456.html

Safety Information

MSDS https://synthonix.com/48456.html
Availability details for
Quantity
  • Availability
  • Add to Cart