SY3H3D681A06 Synthonix Corporation

3-(Pyridin-3-yl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 1126-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/48460.html

Safety Information

MSDS https://synthonix.com/48460.html
Availability details for
Quantity
  • Availability
  • Add to Cart