SY3H3D681A0C Synthonix Corporation

5-Phenyl-1H-1,2,4-triazol-3-amine

  • Cas Number : 4922-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48864.html

Safety Information

MSDS https://synthonix.com/48864.html
Availability details for
Quantity
  • Availability
  • Add to Cart