SY3H3D681A0F Synthonix Corporation

2-Phenyl-2-sulfoacetic acid

  • MDL number No MDL found
  • Cas Number : 41360-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49128.html

Safety Information

MSDS https://synthonix.com/49128.html
Availability details for
Quantity
  • Availability
  • Add to Cart