SY3H3D681A16 Synthonix Corporation

1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 908591-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49557.html

Safety Information

MSDS https://synthonix.com/49557.html
Availability details for
Quantity
  • Availability
  • Add to Cart