SY3H3D681A17 Synthonix Corporation

2-Propylpiperidin-4-one hydrochloride

  • MDL number MFCD11110451
  • Cas Number : 1245645-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49563.html

Safety Information

MSDS https://synthonix.com/49563.html
Availability details for
Quantity
  • Availability
  • Add to Cart