SY3H3D681A1C Synthonix Corporation

(R)-1-(Pyridin-2-yl)propan-1-amine dihydrochloride

  • Cas Number : 1263198-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49666.html

Safety Information

MSDS https://synthonix.com/49666.html
Availability details for
Quantity
  • Availability
  • Add to Cart