SY3H3D681A1F Synthonix Corporation

1-Propylpiperidin-4-amine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1158561-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49682.html

Safety Information

MSDS https://synthonix.com/49682.html
Availability details for
Quantity
  • Availability
  • Add to Cart