SY3H3D681A21 Synthonix Corporation

6-Isopropoxynicotinonitrile

  • MDL number No MDL found
  • Cas Number : 195140-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49720.html

Safety Information

MSDS https://synthonix.com/49720.html
Availability details for
Quantity
  • Availability
  • Add to Cart