SY3H3D681A23 Synthonix Corporation

3-Phenoxypropanal

  • Cas Number : 22409-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link https://synthonix.com/49880.html

Safety Information

MSDS https://synthonix.com/49880.html
Availability details for
Quantity
  • Availability
  • Add to Cart