SY3H3D681A27 Synthonix Corporation

2-(1H-Pyrrolo[3,2-c]pyridin-3-yl)ethanamine

  • MDL number No MDL found
  • Cas Number : 1778-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50001.html

Safety Information

MSDS https://synthonix.com/50001.html
Availability details for
Quantity
  • Availability
  • Add to Cart