SY3H3D681A29 Synthonix Corporation

1-(Pyridin-3-yl)butan-1-one

  • MDL number No MDL found
  • Cas Number : 1701-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50020.html

Safety Information

MSDS https://synthonix.com/50020.html
Availability details for
Quantity
  • Availability
  • Add to Cart